DEEP VENT™ DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0258L
M0258S
CA101227-256EA
727
CAD
CA101227-256
CA101227-258
DEEP VENT™ DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
Deep VentR DNA Polymerase is a high-fidelity thermophilic DNA polymerase
The fidelity of Deep VentR DNA Polymerase is derived in part from an integral 3´→ 5´ proofreading exonuclease activity. Deep VentR is even more stable than VentR at temperatures of 95 to 100°C, with a half-life of 8 hours at 100°C.