DEEP VENT™ (exo-) DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0259L M0259S
CA101227-260EA 727 CAD
CA101227-260 CA101227-262
DEEP VENT™ (exo-) DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Deep VentR (exo-) DNA Polymerase has been genetically engineered to eliminate the 3´→ 5´ proofreading exonuclease activity associated with Deep Vent DNA Polymerase
Deep Vent (exo-) DNA Polymerase is even more stable than VentR® (exo-) DNA Polymerase with a half-life of 23 hours at 95°C and 8 hours at 100°C.