Chitin Resin, New England Biolabs
Supplier: New England Biolabs (NEB)
S6651L S6651S
CA101229-744EA 901 CAD
CA101229-744 CA101229-746
Chitin Resin, New England Biolabs
Protein Linking and Labeling Systems
An affinity matrix for the isolation of target proteins fused to an intein-chitin binding domain fusion.