cAMP-dependent Protein Kinase
Supplier: New England Biolabs (NEB)
P6000L
P6000S
CA101641-522EA
1085
CAD
CA101641-522
CA101641-524
cAMP-dependent Protein Kinase
Enzymes
The catalytic subunit of cAMP-dependent Protein Kinase (PKA) is a serine/threonine protein kinase, which combines, in the absence of cAMP, with the regulatory subunit to form the inactive PKA holoenzyme
Since this is the free catalytic subunit alone, no cAMP is required for activation.