Bst DNA Polymerase Lg Frag, New England Biolabs
Supplier: New England Biolabs (NEB)
M0275L M0275M M0275S
CA101228-330EA 491 CAD
CA101228-330 CA101228-332 CA101228-334
Bst DNA Polymerase Lg Frag, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5´ → 3´ polymerase activity, but lacks 5´ →3´ exonuclease activity.