Bst 2.0 DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0537S M0537L M0537M
CA102500-028EA 123 CAD
CA102500-028 CA102500-030 CA102500-032
Bst 2.0 DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Bst 2.0 DNA Polymerase is an in silico designed homologue of Bacillus stearo thermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment)
Bst 2.0 DNA Polymerase contains 5´→3´ DNA polymerase activity and strong strand displacement activity but lacks 5´→3´ exonuclease activity. Bst 2.0 DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.