beta-Agarase I
Supplier: New England Biolabs (NEB)
M0392L
M0392S
CA101228-394EA
564
CAD
CA101228-394
CA101228-396
beta-Agarase I
Enzymes
β-Agarase cleaves the agarose subunit, unsubstituted neoagarobiose [3,6-anhydro-α-L-galactopyranosyl-1-3-D-galactose] to neoagaro-oligosaccharides
β-Agarase I digests agarose, releasing trapped DNA and producing carbohydrate molecules which can no longer gel. The remaining carbohydrate molecules and β-Agarase I will not, in general, interfere with subsequent DNA manipulations such as restriction endonuclease digestion, ligation and transformation.