BamHI-HF® Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
R3136M R3136T R3136T R3136M
CA101641-130EA 397 CAD
CA101641-130 CA101641-132 101641-130 101641-132
BamHI-HF® Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries the cloned and modified BamHI gene from Bacillus amyloliquefaciens H (ATCC 49763). High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.