Bacteroides Heparinase III
Supplier: New England Biolabs (NEB)
P0737S
P0737L
CA102500-020EA
555
CAD
CA102500-020
CA102500-022
Bacteroides Heparinase III
Enzymes
Bacteroides Heparinase III cloned from Bacteroides eggerthii, also called Heparin Lyase III, is active on both heparin and heparan sulfate
The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm. Bacteroides Heparinase III can cleave the glycosidic bond between hexosamines and either iduronic acid or glucuronic acid residues. Bacteroides Heparinase III is active in the presence of 6-sulfation.