Bacteroides Heparinase II
Supplier: New England Biolabs
P0736S P0736S P0736L P0736L
CA102500-018EA 394 CAD
CA102500-018 102500-016 CA102500-016 102500-018
Bacteroides Heparinase II
Enzymes
Bacteroides Heparinase II cloned from Bacteroides eggerthii, also called Heparin Lyase II, is a low specificity enzyme that is active on both heparin and heparan sulfate
The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm. Bacteroides Heparinase II cleaves the glycosidic bond between N-sulfated and glucuronic or iduronic acid residues. When used alone this enzyme rarely yields complete depolymerization of a polysaccharide chain, however disaccharide analysis is enhanced when used in combination with Heparinase I and III.