Amylose Resin, New England Biolabs
Supplier: New England Biolabs (NEB)
E8021L E8021S
CA101227-542EA 2372 CAD
CA101227-542 CA101227-544
Amylose Resin, New England Biolabs
Protein Purification Protein Purification Resins
Amylose resin is an affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP). It is intended for use in a gravity flow column.
  • Highly specific binding of protein of interest fused to MBP allows for one-step purification
  • MBP is easily removed from the protein of interest by Factor Xa Protease (101228-574, 101228-576)
  • Binding capacity of >4 mg MBP5*-paramyosin ΔSal fusion protein/ml amylose resin
  • Can be used multiple times and can be regenerated