Amylose Resin High Flow, New England Biolabs
Supplier: New England Biolabs (NEB)
E8022L E8022S
CA101227-546EA 3544 CAD
CA101227-546 CA101227-548
Amylose Resin High Flow, New England Biolabs
Protein Purification Protein Purification Resins
Amylose Resin High Flow is a cross-linked affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP)
This rigid matrix can be used in automated chromatography systems. DNASU is a central repository for plasmid clones and collections that may also be helpful.