AgeI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
R3552S R3552S R3552L R3552L
CA101641-200EA 491 CAD
CA101641-200 CA101641-202 101641-200 101641-202
AgeI-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
AgeI-HF® is produced in E. coli from a strain that carries the cloned and modified AgeI gene from Ruegeria gelatinovora (ATCC 25655).
High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.