7-deaza dGTP, New England Biolabs
Supplier: New England Biolabs (NEB)
N0445S N0445L
CA101228-414EA 121 CAD
CA101228-414 CA102500-006
7-deaza dGTP, New England Biolabs
Nucleic Acid Reagents Nucleotides
7-deaza-dGTP (7-deaza-2´-deoxyguanosine 5´-triphosphate) contains a 5 mM solution of 7-deaza-dGTP as a dilithium salt
Nucleotide concentration is determined by measurements of absorbance at 257 nm. Supplied in: Milli-Q® water as a lithium salt at (pH 7.0).