DNA Ladder, 100 bp, New England Biolabs
Supplier: New England Biolabs (NEB)
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis.
The digested DNA includes fragments ranging from 100 to 1517 base pairs. The 500 and 1000 base pair bands have increased intensity to serve as reference points. The approximate mass of DNA in each of the bands is provided (assuming a 0.5 μg load) for approximating the mass of DNA in comparably intense samples of similar size.
Delivery information: Supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.