DNA Ladder, 1 kb, New England Biolabs
Supplier: New England Biolabs (NEB)
N3232L N3232S
CA101228-492EA 355 CAD
CA101228-492 CA101228-494
DNA Ladder, 1 kb, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
A number of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis
The digested DNA includes fragments ranging from 0.5-10.0 kilobases (kb). The 3.0 kb fragment has increased intensity to serve as a reference band. The approximate mass of DNA in each of the bands is provided (assuming a 0.5 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.