Fluid-Resistant Barrier Coats
Supplier: CritiCore Protective Wear
89495-718 89495-722 89495-726 89495-720 89495-732 89495-728 89495-724 89495-716 89495-730 89495-734
89495-718EA 63.11 CAD
89495-718 89495-720 89495-730 89495-724 89495-734 89495-722 89495-732 89495-728 89495-716 89495-726
Fluid-Resistant Barrier Coats
Workwear
These specialized lab coats offer excellent fluid resistance while maintaining their breathability.
  • Fluid-resistant, 100% Polyester front and sleeves
  • PolyCotton back panel for ventilation
  • Quick-release snap front for fast removal
  • Safety-sleeve knit cuff
  • Two hip pockets and one breast pocket
These coats are extremely durable and able to hold up to repeated industrial launderings.
Ordering information: Coats in sizes XS to XXL are stocked. For sizes 3XL to 6XL, extended lead time may be required. Contact your Avantor sales representative for more information.