Cartridges and Columns, LiChrospher® 100 LiChroCART®, MilliporeSigma
Supplier: MilliporeSigma
1.50892.0001 1.50942.0001 1.50956.0001 1.50982.0001 1.50892.0001 1.50956.0001 1.50942.0001 1.50982.0001
48219-434EA 1285.05 CAD
48219-434 48219-454 48219-464 48219-614 CA48219-434 CA48219-464 CA48219-454 CA48219-614
Cartridges and Columns, LiChrospher® 100 LiChroCART®, MilliporeSigma
Chromatography Columns
LiChrospher® CN has polar and hydrophobic properties, thus can be used as less polar alternative to LiChrospher® Si 60 in normal phase applications or as less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications.
LiChrospher® RP-8 is a reliable and versatile traditionally produced spherical silica gel carrier with C-8 reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds. This sorbent is packed into a LiChroCART® 125-4 HPLC cartridge. The connection to the HPLC system is accomplished through the use of the manu-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable