MICROFLEX® NeoTouch™ 25-101, Chemical-Resistant Gloves, Neoprene, Ansell
Supplier: Ansell Healthcare
385727 385728 385729 385730 385731
89011-902CS 657.44 CAD
89011-902 89011-904 89011-906 89011-908 89011-910
MICROFLEX® NeoTouch™ 25-101, Chemical-Resistant Gloves, Neoprene, Ansell
Gloves
This glove is made with neoprene, an alternative to natural rubber latex that protects against Type I allergies.
  • Resistant to acids, bases and alcohols
  • Roll-beaded cuff, powder free
Ideal applications include assembly and inspection, lab analysis and technical work, and pharmaceutical manufacturing - especially when resistance to acids is required.
Their exclusive formulation provides superior comfort.
Certifications: This is an exam- and food-grade glove and complies with FDA food handling requirements (21 CFR 177.2600).
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Finish Textured Fingertips