WHEATON® Standard Opening Crimp Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
223682 223682-01 223692 223692-01
25485-018CS 577.27 CAD
25485-018 89235-520 89235-522 89235-524
WHEATON® Standard Opening Crimp Vials, DWK Life Sciences
Vials
Wheaton Standard Opening Crimp Vials are offered in both clear and amber borosilicate Type I glass.
Limited volume inserts and crimp top seals are available separately.
They are packaged in shrink-wrapped packs of 100 vials to reduce contamination from corrugate. This is the most common chromatography vial used in autosamplers.