Drying Tube Adapter, Straight, Glassco
Supplier: Glassco
041.470.02
89426-424EA 57.17 CAD
89426-424
Drying Tube Adapter, Straight, Glassco
Adapters Adapters, Conical Ground Joints
Drying tube adapters are used for keeping a reaction vessel free of moisture.
  • Straight
  • Top [ST] joint
  • Bottom [ST] joint
Adapter is equipped with standard taper joints at top and bottom.