TouchNTuff® DermaShield® 73-701 Cleanroom gloves, polychloroprene, sterile, Ansell
Supplier: Ansell Canada
7370111060 7370111065 7370111075 7370111080 7370111085 7370111070 7370111090
CA10799-038CS 554.21 CAD
CA10799-038 CA10799-040 CA10799-042 CA10799-044 CA10799-046 CA10799-066 CA10808-964
TouchNTuff® DermaShield® 73-701 Cleanroom gloves, polychloroprene, sterile, Ansell
Gloves
Dermashield is made from a proprietary poly-chloroprene blend that is free of latex proteins and accelerators.
  • Cleanroom compatible
  • Straight cuffs
  • Excellent resistance to many chemicals
These sterile gloves are the ideal choice in the prevention of both Type 1 and Type IV allergies associated with natural rubber.
Dermashield 73-701 is clean and sterile and ideally suited for ISO class 5 (Class 100) enviroments. Neoprene composition offers chemical resistance to a broad spectrum of chemicals without compromising comfort and flexibility.
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Finish Smooth, with Bisque Fingertips