WHEATON® MicroLiter Screw-Thread Vials, 10-425, Component Kits, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
10-1010W 10-1010W-A 10-1040W-A 10-1060W-A 10-1240W 10-1260W-A
97046-958PK 98.75 CAD
97046-958 97046-960 97046-964 97046-966 97046-968 97046-970 97046-976 97046-978 97046-982 97046-984 97046-986 97046-988 97046-996 97046-998 97047-000 97047-002 97047-004 97047-006 89217-240
WHEATON® MicroLiter Screw-Thread Vials, 10-425, Component Kits, DWK Life Sciences
Vials
The original wide-opening vial available for chromatography.
  • Wide-opening
  • Multiple configurations available
For applications where the vial remains stationary, this vial is safe to use, even with inserts. MicroLiter stocks all of the popular septa configurations and convenience kits.