Adapter, with 75° Bend, Glassco
Supplier: GLASSCO LABORATORY EQUIPMENT
043.470.03
89426-430EA 72.05 CAD
89426-430
Adapter, with 75° Bend, Glassco
Adapters Adapters, Conical Ground Joints
Adapter is equipped with a standard taper inner joint at each end.
  • 75° angle bend
  • Top [ST] inner joint
  • Bottom [ST] inner joint