Distillation Connecting Adapter, Glassco
Supplier: Glassco
028.470.02 028.470.03 028.470.04
89426-366EA 85.91 CAD
89426-366 89426-368 89426-372
Distillation Connecting Adapter, Glassco
Distillation Components Distillation Adapters
Adapters are equipped with standard taper outer joint at the top, and inner joints on the bottom and sidearm.
  • Top [ST] outer joint
  • Bottom and sidearm inner joints
Adapter is designed for use as a component in atmospheric or vaccum distillations.