Distillation Trap Adapter, Glassco
Supplier: Glassco
030.476.04A
89426-382EA 108.51 CAD
89426-382
Distillation Trap Adapter, Glassco
Distillation Components Distillation Adapters
This trap adapter is placed above the flask to prevent carry-over of reactions which have a tendency to foam.
  • Inner vapor tube
  • Manufactured from borosilicate 3.3 glass
Adapter has a capacity of 100mL.