Vacuum Adapter, with 1:5 Fluoropolymer Resin Stopcock, Glassco
Supplier: Glassco
034.470.02
89426-406EA 125.02 CAD
89426-406
Vacuum Adapter, with 1:5 Fluoropolymer Resin Stopcock, Glassco
Adapters Adapters, Conical Ground Joints
This vacuum adapter features a hose connection and stopcock.
  • Hose connection
  • Fluoropolymer resin stopcock
  • [ST] joint
Adapter is equipped with an 8mm O.D. hose connection and 2mm fluoropolymer resin stopcock.