Vaccum/Argon Adapter (Introgen), Glassco
Supplier: Glassco
020.475.01 020.475.03
89426-350EA 36.3 CAD
89426-350 89426-352
Vaccum/Argon Adapter (Introgen), Glassco
Adapters Adapters, Conical Ground Joints
Adapters feature a straight hose connection at top and male joint.
  • Top hose connection
  • Male [ST] joint
  • Manufactured from borosilicate 3.3 glass.
Use adapter for vacuum or argon applications.