Sand (from Ottawa), VWR Chemicals BDH®
Supplier: VWR International
BDH9274-2.5KG
BDH9274-500G
BDH9274-2.5KGCS
411.71
CAD
BDH9274-2.5KG
BDH9274-500G
Sand (from Ottawa), VWR Chemicals BDH®
Sand
Beige granules
Formula:
SiO₂ Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1427 °C Density: 2.622…2.645 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02100519 CAS Number: 7631-86-9 EINECS: 238-878-4 |
Specification Test Results
Moisture |
|
Particle Size 30-40 mesh |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...