Reaction Flask, with Glass Stopcock, Glassco
Supplier: Glassco
059.468.07
89426-512EA 226.72 CAD
89426-512
Reaction Flask, with Glass Stopcock, Glassco
Reaction Vessels Reaction Vessels, Flask Shape
Flasks feature a 2mm glass stopcock and a spring retention device for smooth turning purpose.