Round Bottom Flask, Angled, Triple Neck, Glassco
Supplier: Glassco
060.470.00 060.470.00A 060.470.01 060.470.04 060.470.04A 060.470.08A 060.470.09 060.470.10 060.470.19
89426-528EA 114.19 CAD
89426-528 89426-530 89426-532 89426-540 89426-542 89426-544 89426-546 89426-548 89426-550
Round Bottom Flask, Angled, Triple Neck, Glassco
Flasks Round Bottom Flasks
Round bottom flask with joints
Heavy wall design. Manufactured from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform wall thickness.