Round Bottom Flask, Double Neck, Glassco
Supplier: Glassco
059.470.00 059.470.00A 059.470.03 059.470.05A 059.470.08 059.470.10 059.470.15
89426-514EA 95.62 CAD
89426-514 89426-516 89426-518 89426-520 89426-522 89426-524 89426-526
Round Bottom Flask, Double Neck, Glassco
Flasks Round Bottom Flasks
Round bottom flask with same sized joints
Heavy wall design. Manufactured from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform wall thickness.