VWR® B.O.D. Bottles, 60 ml
Supplier: VWR International
89429-188 89429-190 89429-192 89429-194 89429-196 89429-208
89429-188CS 1371.94 CAD
89429-188 89429-190 89429-192 89429-194 89429-196 89429-208
VWR® B.O.D. Bottles, 60 ml
Bottles BOD Bottles
Made from borosilicate glass, these bottles can be used for incubating diluted samples of polluted water, industrial wastes and more.
They come with a permanently screened label and are available with glass stoppers.