VWR® Powder Funnels, Wide Stem
Supplier: VWR International
H14660-0065 H14660-0150 H14660-0080 H14660-0100
16126-912CS 308.73 CAD
16126-912 16126-916 10033-456 10033-458
VWR® Powder Funnels, Wide Stem
Funnels Powder Funnels
Polypropylene funnels are steam autoclavable at 121°C (250°F).
  • Wide stem minimizes bridging of powder
  • 60° cone angle
  • Polypropylene construction