MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
Supplier: MilliporeSigma
1.10020.0001 1.10020.0002 1.10092.0021
CA1.10020.0001EA 179 CAD
CA1.10020.0001 CA1.10020.0002 CA1.10092.0021
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
Environmental Test Kits
The nitrate test can be used for monitoring of drinking water, process water and wastewater, as well as aquarium water where limits can quickly be exceeded
MilliporeSigma # 1.10020.0001 replaces and is identical to MilliporeSigma # 10020-1
It can also be used for determine nitrate in food e.g. potatoes.
Accessories information: Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.