pH-Indicator Paper Litmus Roll, MilliporeSigma
Supplier: MilliporeSigma
1.09486.0003 1.09489.0003 1.09489.0003 1.09486.0003
CA1.09489.0003CS 12327.21 CAD
EM1.09489.0003 EM1.09486.0003 CA1.09489.0003 CA1.09486.0003
pH-Indicator Paper Litmus Roll, MilliporeSigma
Test Papers pH Indicator Papers and Test Strips
Roll with color scale Ph Eur.
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.