PTFE Covered Stainless Steel Stir Shaft with 4-Blade 45° Agitator, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
13852-10 13852-15 13852-19
89428-024EA 1120.3 CAD
89428-024 89428-026 89428-028
PTFE Covered Stainless Steel Stir Shaft with 4-Blade 45° Agitator, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
This PTFE covered stainless steel shaft features a solid PTFE, 4-blade screw propeller
Blades are pitched 45 degrees. To provide a better grip by the stir motor chuck, 50mm of stainless steel shaft is left exposed.
Order Now
SPECIFICATIONS
Environmentally Preferable