Flowmeters, CO₂, AIMS
Supplier: Aims
SC100 LC500 XL900
52428-576EA 1241.28 CAD
52428-576 52428-578 52428-580
Flowmeters, CO₂, AIMS
Flow Meters
Calibrated to accurately deliver CO2 to the chamber
It is easy to connect to any chamber and has a small profile. Three-way ball valve offers quick chamber fill when needed. Easy to set flow rate and is wall mountable.