Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm, Ace Glass Incorporated
Supplier: Ace Glass
7821-01 7821-03 7821-05 7821-07
89427-992EA 343.14 CAD
89427-992 89427-994 89427-996 89427-998
Vacuum Storage Tube with Hi-Vac Top Valve and Outer Standard Taper Joint Sidearm, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate glass vacuum storage tube features an 0-4mm Hi-Vac valve top with a standard taper jointed sidearm
The valved top enables precise flow control and the standard taper, outer joint facilitates connections to Schlenk lines.
Order Now
SPECIFICATIONS
Environmentally Preferable