General Purpose Water Baths, Boekel Scientific
Supplier: Boekel
290200 290300 290100
97004-338EA 3826.37 CAD
97004-338 97004-342 97004-344
General Purpose Water Baths, Boekel Scientific
Baths Water Baths, Unstirred
The Boekel Scientific family of Water Baths offers a variety of sizes to meet all of your needs and is ideal for general laboratory applications
Equipped with an adjustable over temperature safety cutout which sounds an audible alarm and shuts the heater off helps protect your samples while the recessed control panel helps protect against spills. Also featured is a seamless passivated tank to prevent against rust and corrosion.
The space saving design allows for the most efficient use of your bench space and the Microprocessor Controller makes for easy temperature selection, rapid heat-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature range Ambient +5°C to 100°C
Timer Settable to 99 hrs, 59 min or "OFF"