Glass 'Y' Shaped Vacuum Adapter, with One Inner and Two Outer Standard Taper Joints, Ace Glass Incorporated
Supplier: Ace Glass
5239-02 5239-04
89427-898EA 268.8 CAD
89427-898 89427-900
Glass 'Y' Shaped Vacuum Adapter, with One Inner and Two Outer Standard Taper Joints, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Glass "Y" shaped three-way vacuum adpater.
  • Two outer [ST] joints
  • One inner [ST] joint
  • Great for adapting flasks and other Schlenk line components
Adapter features two outer standard taper joints, and one inner standard taper joint. Great for adapting flasks and other Schlenk line components. All three joints are the same size. Fabricated from borosilicate glass.