Adjustable Height Tables, Diversified Woodcrafts
Supplier: Diversified Woodcrafts
P8304K P8204K P8201K P8101K P8104K P8604K P8144K P8301K P8601K P8141K
89375-184EA 5370.91 CAD
89375-186 89375-174 89375-196 89375-184 89375-278 89375-178 89375-190 89375-282 89375-192 89375-180
Adjustable Height Tables, Diversified Woodcrafts
Furniture Tables and Workbenches
A perfect solution for ADA requirements that can adjust in 2 cm increments from 27.75" to 39" (42" with 3" spacer provided).
  • Can also be adjusted for use as an incline surface for physics
  • Solid 4.5" oak aprons are attached to your choice of three top options: 1.25" plastic laminate, 1.25" ChemGuard, and 0.75" phenolic
The wood is finished with a chemical resistant, earth-friendly UV finish. The table has a weight capacity of 500 lbs.
Ordering information: Optional casters sold separately.
Caution: Phenolic resin tops are heavier and may require two people to adjust the height.