Lab Armor® StayTemp™ Trays, Shel Lab
Supplier: Sheldon Manufacturing
39956-050 39956-001 39956-003 39956-003
89409-262EA 571.75 CAD
89409-262 89409-264 89409-266 490014-034
Lab Armor® StayTemp™ Trays, Shel Lab
Trays/Dishes/Pots Laboratory Trays/Dishes
StayTemp™ equipment trays are an exciting new upgrade that keeps samples and reagents in your refrigerator, incubator, and laboratory ovens at temperature despite normal hourly fluctuations in air temperature
Stainless steel construction. For use in refrigerators, incubators and ovens. Does not include a lid. Hand wash, autoclave, towel dry.
You no longer have to worry about how many times the doors were opened or whether or not your samples experience a defrost cycle.
Order Now
SPECIFICATIONS
Environmentally Preferable