Lab Armor® Beads, Sheldon Manufacturing
Supplier: Sheldon Manufacturing
42370-004 42370-008 42370-002 42370-750
89409-244EA 1879.41 CAD
89409-244 89409-246 89409-240 89409-242
Lab Armor® Beads, Sheldon Manufacturing
Beads Metallic Beads
Recyclable Lab Armor Beads eliminate water bath contamination, maintenance, and harmful germicides
Lab Armor Beads will help you save time and money. No more hassling with emptying, cleaning and refilling water baths. The bath stays on so there is no need to plan around warm-up times. Lab Armor Beads eliminate the worry about burnout because there is no water to evaporate.
Other benefits of using Lab Armor Beads include cleaner working environment, greater organization capabilities as samples are held in place, and environmental friendly, non-toxic, non-vaporizing material. No daily requirement for biocide use-no gray water. Improves energy efficiency of standard water baths by over 50%.
Dry metallic beads that replace water in baths give old equipment a cool, shiny, state-of-the-art upgrade.
Weight: 3.65 lbs./L
Working temperature range: −80...180 °C
Size: ~5-8 mm x 1,2 mm (ØxH)