C-MAG HP 4 IKATHERM® Hot Plate, IKA® Works
Supplier: IKA Works
0003581601 3581600
89495-336EA 1121.8 CAD
89495-336 89495-338
C-MAG HP 4 IKATHERM® Hot Plate, IKA® Works
Heaters Hot Plates
Newly designed hotplate constructed of ceramic glass to offer excellent chemical resistance.
Standard features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, exact temperature setting via digital display (LED), a digital error code display, and an elevated control panel for protection against liquid leaks.
Units meet IP 21 protection class according to DIN EN 60529.
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with a three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Accessories are sold separately.