Platinum-Cured Silicone Pump Tubing on a Spool, NewAge Industries
Supplier: NewAge Industries
CA7021019 CA7021020 CA7021021 CA7021022 89511-340
52428-898EA 3209.53 CAD
52428-898 52428-900 52428-902 52428-904 89511-340
Platinum-Cured Silicone Pump Tubing on a Spool, NewAge Industries
Tubing Transfer Tubing
Extremely flexible tubing is specially formulated to withstand repeated compression and release for consistent, reliable performance in peristaltic pumps.
  • Sterilizable by autoclave and gamma radiation
  • Imparts no taste or odor to critical stream
Low-volatile grade, platinum-cured silicone tubing is produced in a certified ISO Class 6 (FED-STD-209E Class 1000/M4.5) cleanroom for use in critical pharmaceutical, biomedical, laboratory, food, beverage, and fragrance applications. Designed to maintain a long service life, this tubing features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
Tubing has undergone extensive physical, chemical, and biological testing and meets USP Class VI and FDA CFR 177.2600 standards; compound meets ISO 10993 standards.
Certifications: Certified free of animal-derived components.
Ordering information: Supplied in (100' to 500') lengths. Sealed in polyethylene bags to ensure cleanliness. Quality control and lot traceability identification on bags and boxes. Complete validation package available upon request.
Caution: Note: Not intended for implantation, continuous stream applications, or elevated pressure levels.