Glass 'Y' Shaped Vacuum Adapter, with One Outer and Two Inner Standard Taper Joints, Ace Glass Incorporated
Supplier: Ace Glass
5238-01 5238-03
89427-894EA 228.44 CAD
89427-894 89427-896
Glass 'Y' Shaped Vacuum Adapter, with One Outer and Two Inner Standard Taper Joints, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Glass "Y" shaped, three-way vacuum adpater.
  • One outer joint
  • Two inner [ST] joints
  • Great for adapting flasks and other Schlenk line components
Adapter features one outer joint and two inner standard taper joints. It is great for adapting flasks and other Schlenk line components. All three joints are the same size on each adapter. Fabricated from borosilicate glass.