Microfuge® 20 Series IVD Microcentrifuges, Beckman Coulter®
Supplier: Beckman Coulter
B30139 B30147 B30147 B30139 B30148 B30138 B30149 B30148 B30137 B30149 B30137 B30138
CA89429-548EA 15755.62 CAD
CA89429-548 89429-540 CA89429-550 89429-550 89429-544 CA89429-542 CA89429-540 89429-542 CA89429-546 89429-548 89429-546 CA89429-544
Microfuge® 20 Series IVD Microcentrifuges, Beckman Coulter®
Centrifuges Microcentrifuges
Fast and versatile, the high-performance Microfuge® 20 Series are precisely designed to meet a wide variety of application needs.
  • Applications: Nucleic acid and protein preparation, Pelleting, Extractions, Purifications, Concentrations, and more
  • Available in Refrigerated and Non-Refrigerated Designs
  • Fast, Sample Processing at Maximum Speeds of up to 15,000 rpm
  • Easy-to-Use Interface Displays Speed and RCF
  • Energy-Efficient
The Microfuge 20 Series offers four different rotors to choose from, including a BioCertified option
Paired with Beckman Coulter Microfuge Series rotors, this mini-centrifuge series offers you all the flexibility you need for your separation protocols.
The four fixed-angle rotors are FA361.5 (2 mL, 36 x 1.5"), FA241.5 (2 mL, 24 x 1.5"), FA241.5P (2 mL, 24 x 1.5"), and FA4x8.2P (2 mL, 4 x 8") PCR tube strip. The Microfuge 20 Series offers a maximum RCF of 20,627 xg with F241.5 and F241.5P rotors and quick acceleration. The Microfuge 20 and Microfuge 20R accelerate in as little as 11 seconds.
Certifications: IVD, TUV Certified for applicable U.S and Canadian standards.
Ordering information: Six configurations are available: Microfuge 20 with FA241.5P rotor, Microfuge 20 with FA241.5 rotor, Microfuge 20 with FA361.5 rotor, Microfuge 20R with FA241.5P rotor, Microfuge 20R with FA241.5 rotor and Microfuge 20R with FA361.5 rotor.
Supplied with manufacturer's shipped two-year warranty.