5215 CO₂ Air Jacketed Infrared Incubators, Shel Lab
Supplier: Sheldon Manufacturing
SCO5A SCO10A
89409-296EA 19175.7 CAD
89409-296 89409-298
5215 CO₂ Air Jacketed Infrared Incubators, Shel Lab
Incubators CO₂ Incubators
Air jacket type Model 5215 was designed to minimize contamination and be operational within hours of installation.
  • HEPA Filter
  • Infrared CO₂ Sensor
  • Heated Outer Doors
  • Independent Overtemperature Protection
  • Quick-Clean Shelf System
  • Access ports: 1,5''
This slim, lightweight incubator is easily repositioned with minimal downtime and is well-suited for multiple users. Three temperature control settings (main chamber, external door, and front liner) minimize condensation and yield precise temperature uniformity with no unwelcome temperature gradients.Our unique HEPA filtration system removes 99.97% of all airborne microbes and isolated particulates 0.3 microns or larger. The patented copper housing around the filter is designed to destroy microbes entrapped in the filter. Additional anti-contamination features include a stainless steel chamber with easy-to-clean coved corners.
A separate digital keypad with large, bright LEDs (for CO₂ and temperature) is highly visible on the front panel. The audible alarms can be muted easily, and the redundant safety system for temperature protects samples from overheating. In addition, the CO₂ gas supply is protected by a PID with an infrared sensor that is extremely quick to recover, a door switch that automatically turns off the gas when the door is opened, and a sample port on the side to help minimize waste during CO₂ calibration.
Ordering information: 5 year limited parts warranty. 7 year warranty on IR sensor.