ART® Barrier Reload System, Thermo Scientific
Supplier: Thermo Scientific
2139-RT 2139-05-RT 2149P-RT 2149P-05-RT 2069-RT 2069-05-RT 2070-RT 2070-05-RT 2179-RT 2179-05-RT
89413-184CS 1474.17 CAD
89413-184 89413-186 89413-212 89413-214 89413-232 89413-234 89413-266 89413-270 89413-292 89413-294
ART® Barrier Reload System, Thermo Scientific
Pipette Tips
ART® portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in the laboratory.
Due to the established level of protection, researchers have made ART® self-sealing barrier tips their first choice in sample protection. ART® barrier tips provide 100% security against aerosols and liquid contamination. The space saving, stackable tip packaging allows for tips to pass through offset holes in lower insert. The easy to use blister sleeve guide rails align tip inserts for a smooth transfer into the rack. Supplied in sterile packaging, the outer sleeve protects tips from exterior contamination while refilling inserts. The environmentally-friendly tip packaging option reduces plastic waste. The rigid insert design provides reliable tip loading into multichannel devices. All tips are traceable, with lot number and description printed on every insert.
Renowned for reliable quality, the ART® series of pipette tips is now offering a complete selection of Barrier, Non-Filter, and Specialty tips. All formats deliver proven quality, performance, and reliability, backed by the highest Thermo Scientific standards and worldwide support.
Certifications: RNase-, DNase-, DNA-, ATP-, and pyrogen-free.
Ordering information: An empty rack is required for Reload Tower use.